Assay Details
| Gene Symbol: NEWGENE_1359502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237685.2 | XP_006237747.1 | 9 - 10 | 2432 | 106 | |
| XM_006237686.2 | XP_006237748.1 | 9 - 10 | 1502 | 106 | ||
| XM_006237687.2 | XP_006237749.1 | 8 - 9 | 1424 | 106 | ||
| XM_008763455.1 | XP_008761677.1 | 10 - 11 | 2510 | 106 | ||
| XM_008763456.1 | XP_008761678.1 | 10 - 11 | 3884 | 106 | ||
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014061.1 | NP_001014083.1 | 1097 | 106 | ||
| GenBank mRNA | BC079135.1 | - | 1097 | 7127603 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P884119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24924 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P884119 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P884119 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P884119 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P330908 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P884119 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P825542 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38538 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16845 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79351 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10210 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61552 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69977 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56802 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31603 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29080 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40622 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30212 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21800 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67895 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10859745 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859746 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859743 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859740 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859744 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859747 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859742 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10859738 | - | - |
| Rat Genome 230 2.0 Array | 1386755_at | - | - |
| Rat Genome 230 2.0 Array | 1383594_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386755_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381991_at | - | - |
| Rat Genome 230 2.0 Array | 1381991_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383594_at | - | - |
| Affymetrix HT Rat Focus Array | 1383594_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383594_at | - | - |
| Affymetrix HT Rat Focus Array | 1381991_at | - | - |
| Affymetrix HT Rat Focus Array | 1386755_at | - | - |
| Affymetrix HT Rat Focus Array | 1381991_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386755_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400014941.rn.1 | TC0400002075.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400014943.rn.1 | TC0400002075.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400014942.rn.1 | TC0400002075.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400014944.rn.1 | TC0400002075.rn.1 | RTA TC |
| Rat Expression Set 230 | 1381991_at | - | - |
| Rat Expression Set 230 | 1383594_PM_at | - | - |
| Rat Expression Set 230 | 1386755_PM_at | - | - |
| Rat Expression Set 230 | 1383594_at | - | - |
| Rat Expression Set 230 | 1381991_PM_at | - | - |
| Rat Expression Set 230 | 1386755_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859738 | - | - |
| Rat Genome U34 Set | rc_AI170070_at | - | - |
| Rat Genome U34 Set | rc_AI177144_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21969321 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650500 | - | - |

NEWGENE_1359502
Genomic Map