Assay Details
| Gene Symbol: NEWGENE_1359502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237685.2 | XP_006237747.1 | 1 - 2 | 1564 | 106 | |
| XM_008763455.1 | XP_008761677.1 | 1 - 2 | 1564 | 106 | ||
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014061.1 | NP_001014083.1 | 151 | 106 | ||
| GenBank mRNA | BC079135.1 | - | 151 | 7127603 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079135.1, NM_001014061.1, XM_006237685.2, XM_008763455.1 transcripts.
Set Membership: |
|

NEWGENE_1359502
Genomic Map