Assay Details
| Gene Symbol: NEWGENE_1359502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237686.2 | XP_006237748.1 | 5 - 6 | 1155 | 70 | |
| XM_008763455.1 | XP_008761677.1 | 6 - 7 | 2163 | 70 | ||
| XM_008763456.1 | XP_008761678.1 | 6 - 7 | 3537 | 70 | ||
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014061.1 | NP_001014083.1 | 750 | 70 | ||
| GenBank mRNA | BC079135.1 | - | 750 | 7127603 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

NEWGENE_1359502
Genomic Map