Assay Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100489.1 | NP_001093959.1 | 2 - 3 | 279 | 72 | |
| XM_006237179.1 | XP_006237241.1 | 1 - 2 | 157 | 72 | ||
| GenBank mRNA | BC161860.1 | - | 422 | 7096336 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wnt5b
Genomic Map