Assay Details
| Gene Symbol: Ppap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231949.2 | XP_006232011.1 | 1 - 2 | 519 | 63 | |
| GenBank mRNA | AF503609.1 | - | 1 - 2 | 57 | 63 | |
More Information
Set Membership: |
|

Ppap2a
Genomic Map