Assay Details
| Gene Symbol: Gpr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138891.1 | NP_620246.1 | 1 - 2 | 1518 | 98 | |
| GenBank mRNA | AY030276.1 | - | 1 - 2 | 1518 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr149
Genomic Map