Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014253.2 | NP_001014275.2 | 2 - 3 | 308 | 157 | |
| GenBank mRNA | AY995234.1 | - | 2 - 3 | 308 | 157 | |
| BC086953.1 | - | 2 - 3 | 202 | 7321890 | 157 | |
| FQ211813.1 | - | 326 | 157 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Selt
Genomic Map