Assay Details
| Gene Symbol: Aadac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020538.1 | NP_065413.1 | 2 - 3 | 384 | 157 | |
| GenBank mRNA | AF182426.1 | - | 2 - 3 | 384 | 157 | |
| BC088143.1 | - | 2 - 3 | 385 | 7323135 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aadac
Genomic Map