Assay Details
| Gene Symbol: Spg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106433.2 | NP_001099903.2 | 2 - 3 | 889 | 129 | |
| XM_006232365.2 | XP_006232427.1 | 2 - 3 | 1217 | 129 | ||
| XM_006232366.2 | XP_006232428.1 | 2 - 3 | 1208 | 129 | ||
| XM_006232367.2 | XP_006232429.1 | 2 - 3 | 1006 | 129 | ||
More Information
Set Membership: |
|

Spg20
Genomic Map