Assay Details
| Gene Symbol: Vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031353.1 | NP_112643.1 | 2 - 3 | 129 | 77 | |
| XM_006246346.1 | XP_006246408.1 | 1 - 2 | 102 | 77 | ||
| GenBank mRNA | AB039662.1 | - | 2 - 3 | 129 | 77 | |
| AF048828.1 | - | 2 - 3 | 119 | 77 | ||
| AF268467.1 | - | 1 - 2 | 63 | 77 | ||
| BC060558.1 | - | 2 - 3 | 106 | 77 | ||
| BC072484.1 | - | 2 - 3 | 113 | 7105146 | 77 | |
| BC087573.1 | - | 2 - 3 | 108 | 7304939 | 77 | |
| BC087657.1 | - | 2 - 3 | 85 | 7312189 | 77 | |
| BC104684.1 | - | 2 - 3 | 99 | 7462139 | 77 | |
| BC127491.1 | - | 2 - 3 | 85 | 8370617 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vdac1
Genomic Map