Assay Details
| Gene Symbol: Cxcl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017478.1 | NP_001017478.1 | 1 - 2 | 355 | 95 | |
| GenBank mRNA | BC079312.1 | - | 1 - 2 | 355 | 7134808 | 95 |
| DQ025528.1 | - | 1 - 2 | 631 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl16
Genomic Map