Assay Details
| Gene Symbol: Cxcl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017478.1 | NP_001017478.1 | 2 - 3 | 491 | 70 | |
| GenBank mRNA | BC079312.1 | - | 2 - 3 | 491 | 7134808 | 70 |
| DQ025528.1 | - | 2 - 3 | 767 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl16
Genomic Map