Assay Details
| Gene Symbol: Krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY464140.1 | - | 1 - 2 | 488 | 91 | |
| BC088424.1 | - | 1 - 2 | 484 | 91 | ||
| BC126075.1 | - | 1 - 2 | 479 | 7442082 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt19
Genomic Map