Assay Details
| Gene Symbol: Krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY464140.1 | - | 3 - 4 | 729 | 117 | |
| BC088424.1 | - | 3 - 4 | 725 | 117 | ||
| BC126075.1 | - | 3 - 4 | 720 | 7442082 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt19
Genomic Map