Assay Details
| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053639.2 | NP_446091.2 | 3 - 4 | 255 | 66 | |
| XM_006246219.2 | XP_006246281.1 | 3 - 4 | 448 | 66 | ||
| XM_008767655.1 | XP_008765877.1 | 4 - 5 | 1276 | 66 | ||
| GenBank mRNA | AB048790.2 | - | 3 - 4 | 255 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ltc4s
Genomic Map