Assay Details
| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021581.1 | NP_067592.1 | 2 - 3 | 557 | 94 | |
| XM_006247414.2 | XP_006247476.1 | 2 - 3 | 861 | 94 | ||
| GenBank mRNA | X65454.1 | - | 2 - 3 | 557 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

P3h4
Genomic Map