Assay Details
| Gene Symbol: Tvp23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107568.3 | NP_001101038.2 | 2 - 3 | 133 | 71 | |
| GenBank mRNA | BC168178.1 | - | 2 - 3 | 131 | 71 | |
| CK483802.1 | - | 2 - 3 | 133 | 71 | ||
| FQ212667.1 | - | 2 - 3 | 169 | 71 | ||
| FQ219631.1 | - | 2 - 3 | 134 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tvp23b
Genomic Map