Assay Details
| Gene Symbol: Zfp711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002730265.4 | XP_002730311.1 | 3 - 4 | 977 | 131 | |
| XM_008773361.1 | XP_008771583.1 | 5 - 6 | 2050 | 131 | ||
| XM_008773362.1 | XP_008771584.1 | 4 - 5 | 1796 | 131 | ||
More Information
Set Membership: |
|

Zfp711
Genomic Map