Assay Details
| Gene Symbol: Npl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013984.1 | NP_001014006.1 | 9 - 10 | 716 | 78 | |
| XM_006249997.2 | XP_006250059.1 | 8 - 9 | 771 | 78 | ||
| GenBank mRNA | BC082004.1 | - | 9 - 10 | 716 | 7131519 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P777173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163006 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P777173 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P777173 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P777173 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P777173 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P309160 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71876 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70343 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14209 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97987 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10768733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768737 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768730 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768735 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10768726 | - | - |
| Rat Genome 230 2.0 Array | 1382874_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382874_at | - | - |
| Affymetrix HT Rat Focus Array | 1382874_at | - | - |
| Affymetrix HT Rat Focus Array | 1382874_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300011364.rn.1 | TC1300001649.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382874_at | - | - |
| Rat Expression Set 230 | 1382874_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768726 | - | - |
| Rat Genome U34 Set | rc_AA956832_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21601055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363958 | - | - |

Npl
Genomic Map