Assay Details
| Gene Symbol: Gas5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002704.1 | 6 - 7 | 189 | 81 | ||
| GenBank mRNA | FQ233900.1 | - | 6 - 7 | 216 | 81 | |
| U77829.1 | - | 6 - 7 | 189 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 37 | Zbtb37 | RGD1308731 | -2.0kb | Anti-sense | Chr. 13 - 78836491 - 78852211 on NCBI Build Rnor_6.0 |
| Downstream | aspartyl-tRNA synthetase 2 (mitochondrial) | Dars2 | RGD1308286 | 3.0kb | Anti-sense | Chr. 13 - 78857638 - 78885464 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

Gas5
Genomic Map