Assay Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113789.1 | NP_001107261.1 | 5 - 6 | 861 | 60 | |
| NM_001143756.1 | NP_001137228.1 | 5 - 6 | 700 | 60 | ||
| XM_006230495.2 | XP_006230557.1 | 5 - 6 | 774 | 60 | ||
| XM_008760008.1 | XP_008758230.1 | 5 - 6 | 1602 | 60 | ||
| XM_008760009.1 | XP_008758231.1 | 5 - 6 | 741 | 60 | ||
| GenBank mRNA | BC158629.1 | - | 4 - 5 | 598 | 7456037 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc27
Genomic Map