Assay Details
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106320.2 | NP_001099790.2 | 21 - 22 | 3038 | 70 | |
| XM_006230715.2 | XP_006230777.1 | 25 - 26 | 3234 | 70 | ||
| XM_006230717.2 | XP_006230779.1 | 24 - 25 | 3204 | 70 | ||
| XM_006230718.2 | XP_006230780.1 | 25 - 26 | 3234 | 70 | ||
| XM_006230719.2 | XP_006230781.1 | 24 - 25 | 3171 | 70 | ||
| XM_006230720.2 | XP_006230782.1 | 24 - 25 | 3159 | 70 | ||
| XM_006230721.2 | XP_006230783.1 | 23 - 24 | 3144 | 70 | ||
| XM_006230722.2 | XP_006230784.1 | 21 - 22 | 3039 | 70 | ||
| XM_006230723.2 | XP_006230785.1 | 21 - 22 | 3039 | 70 | ||
| XM_006230724.2 | XP_006230786.1 | 23 - 24 | 3096 | 70 | ||
| Gene Symbol: LOC103689951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749028.3 | XP_003749076.2 | 4 - 5 | 540 | 70 | |
| XM_006230626.2 | XP_006230688.1 | 3 - 4 | 513 | 70 | ||
| XM_006230627.2 | XP_006230689.1 | 4 - 5 | 541 | 70 | ||
| XM_006230628.2 | XP_006230690.1 | 3 - 4 | 476 | 70 | ||
| XM_006230629.2 | XP_006230691.1 | 2 - 3 | 447 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppfia1
Genomic Map