Assay Details
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230715.2 | XP_006230777.1 | 30 - 31 | 3812 | 72 | |
| XM_006230717.2 | XP_006230779.1 | 29 - 30 | 3782 | 72 | ||
| XM_006230719.2 | XP_006230781.1 | 29 - 30 | 3749 | 72 | ||
| XM_006230720.2 | XP_006230782.1 | 29 - 30 | 3737 | 72 | ||
| XM_006230721.2 | XP_006230783.1 | 28 - 29 | 3722 | 72 | ||
| XM_006230722.2 | XP_006230784.1 | 26 - 27 | 3617 | 72 | ||
| XM_006230724.2 | XP_006230786.1 | 28 - 29 | 3674 | 72 | ||
| Gene Symbol: LOC103689951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749028.3 | XP_003749076.2 | 9 - 10 | 1118 | 72 | |
| XM_006230626.2 | XP_006230688.1 | 8 - 9 | 1091 | 72 | ||
| XM_006230628.2 | XP_006230690.1 | 8 - 9 | 1054 | 72 | ||
| XM_006230629.2 | XP_006230691.1 | 7 - 8 | 1025 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ppfia1
Genomic Map