Assay Details
| Gene Symbol: Rhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106323.1 | NP_001099793.1 | 2 - 3 | 314 | 77 | |
| GenBank mRNA | FQ219373.1 | - | 2 - 3 | 263 | 77 | |
| FQ229623.1 | - | 2 - 3 | 263 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rhod
Genomic Map