Assay Details
| Gene Symbol: Rhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106323.1 | NP_001099793.1 | 3 - 4 | 422 | 103 | |
| GenBank mRNA | FQ229623.1 | - | 3 - 4 | 371 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhod
Genomic Map