Assay Details
| Gene Symbol: Colec11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239960.2 | XP_006240022.1 | 4 - 5 | 528 | 64 | |
| XM_006239961.1 | XP_006240023.1 | 4 - 5 | 321 | 64 | ||
| XM_006239962.2 | XP_006240024.1 | 4 - 5 | 527 | 64 | ||
| GenBank mRNA | BC158664.1 | - | 324 | 64 | ||
More Information
Set Membership: |
|

Colec11
Genomic Map