Assay Details
| Gene Symbol: Fkbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105922.1 | NP_001099392.1 | 3 - 4 | 302 | 64 | |
| XM_006249131.2 | XP_006249193.1 | 3 - 4 | 321 | 64 | ||
| XM_008769130.1 | XP_008767352.1 | 3 - 4 | 321 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp6
Genomic Map