Assay Details
| Gene Symbol: Ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108331.2 | NP_001101801.2 | 3 - 4 | 371 | 84 | |
| XM_006249179.2 | XP_006249241.1 | 3 - 4 | 400 | 84 | ||
| GenBank mRNA | BC169070.1 | - | 3 - 4 | 315 | 5598714 | 84 |
| FQ230081.1 | - | 2 - 3 | 235 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap1s1
Genomic Map