Assay Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004204.2 | NP_001004204.1 | 7 - 8 | 1156 | 72 | |
| XM_006249230.2 | XP_006249292.1 | 4 - 5 | 858 | 72 | ||
| XM_006249231.2 | XP_006249293.1 | 4 - 5 | 779 | 72 | ||
| GenBank mRNA | BC079161.1 | - | 4 - 5 | 830 | 7129411 | 72 |
| BC090318.1 | - | 7 - 8 | 1156 | 7113430 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem248
Genomic Map