Assay Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004267.1 | NP_001004267.1 | 8 - 9 | 808 | 73 | |
| GenBank mRNA | BC079127.1 | - | 8 - 9 | 808 | 7127112 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P233753 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P522477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120647 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P233753 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P522477 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P522477 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P233753 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P522477 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P233753 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P233753 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P522477 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54159 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10861073 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861072 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861070 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861071 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861068 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861076 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861075 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861069 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10861066 | - | - |
| Rat Genome 230 2.0 Array | 1383731_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392486_at | - | - |
| Rat Genome 230 2.0 Array | 1383731_at | - | - |
| Rat Genome 230 2.0 Array | 1395920_at | - | - |
| Rat Genome 230 2.0 Array | 1392486_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1395920_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395920_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383731_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395920_at | - | - |
| Affymetrix HT Rat Focus Array | 1392486_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392486_at | - | - |
| Affymetrix HT Rat Focus Array | 1383731_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400002817.rn.1 | TC0400000382.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400002818.rn.1 | TC0400000382.rn.1 | RTA TC |
| Rat Expression Set 230 | 1383731_at | - | - |
| Rat Expression Set 230 | 1392486_PM_at | - | - |
| Rat Expression Set 230 | 1392486_at | - | - |
| Rat Expression Set 230 | 1383731_PM_at | - | - |
| Rat Expression Set 230 | 1395920_at | - | - |
| Rat Expression Set 230 | 1395920_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861066 | - | - |
| Rat Genome U34 Set | rc_AA818065_at | - | - |
| Rat Genome U34 Set | rc_AI178159_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21679739 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371474 | - | - |
| RatRef-12 Expression BeadChip | ILMN_50489 | - | - |

Tmem106b
Genomic Map