Assay Details
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105668.1 | NP_001099138.1 | 3 - 4 | 571 | 115 | |
| XM_006236102.2 | XP_006236164.1 | 3 - 4 | 847 | 115 | ||
| XM_006236103.2 | XP_006236165.1 | 3 - 4 | 876 | 115 | ||
| GenBank mRNA | CV106654.1 | - | 372 | 115 | ||
| DN932316.1 | - | 3 - 4 | 595 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdfic
Genomic Map