Assay Details
| Gene Symbol: Mob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109159.1 | NP_001102629.1 | 424 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P830152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132332 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P830152 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P830152 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P830152 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P529743 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98890 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10726928 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726927 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10726924 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10726924 | - | - |
| Rat Genome 230 2.0 Array | 1389614_at | - | - |
| Rat Genome 230 2.0 Array | 1389614_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100055003.rn.1 | TC0100006963.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100054995.rn.1 | TC0100006963.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100055004.rn.1 | TC0100006963.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100055005.rn.1 | TC0100006963.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100054997.rn.1 | TC0100006963.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100055002.rn.1 | TC0100006963.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389614_PM_at | - | - |
| Rat Expression Set 230 | 1389614_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10726924 | - | - |
| Rat Genome U34 Set | rc_AI172472_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21730305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366200 | - | - |

Mob2
Genomic Map