Assay Details
| Gene Symbol: Ms4a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231110.2 | XP_006231172.1 | 5 - 6 | 994 | 137 | |
| XM_215145.8 | XP_215145.5 | 5 - 6 | 724 | 137 | ||
More Information
Set Membership: |
|

Ms4a6b
Genomic Map