Assay Details
| Gene Symbol: Mrpl49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047883.1 | NP_001041348.1 | 3 - 4 | 392 | 98 | |
| GenBank mRNA | AY325168.1 | - | 3 - 4 | 392 | 98 | |
| FQ209431.1 | - | 357 | 98 | |||
| FQ212546.1 | - | 371 | 98 | |||
| FQ235268.1 | - | 371 | 98 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl49
Genomic Map