Assay Details
| Gene Symbol: Bccip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108505.2 | NP_001101975.2 | 1 - 2 | 194 | 144 | |
| GenBank mRNA | FQ227185.1 | - | 1 - 2 | 185 | 144 | |
| FQ228402.1 | - | 1 - 2 | 192 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bccip
Genomic Map