Assay Details
| Gene Symbol: Prdx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053610.1 | NP_446062.1 | 4 - 5 | 512 | 98 | |
| GenBank mRNA | AF110732.1 | - | 4 - 5 | 512 | 98 | |
| BC078771.1 | - | 4 - 5 | 488 | 7193191 | 98 | |
| FQ210686.1 | - | 4 - 5 | 443 | 98 | ||
| FQ223238.1 | - | 4 - 5 | 440 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdx5
Genomic Map