Assay Details
| Gene Symbol: Kitlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021843.4 | NP_068615.1 | 2 - 3 | 306 | 104 | |
| NM_021844.2 | NP_068616.1 | 2 - 3 | 306 | 104 | ||
| XM_008765339.1 | XP_008763561.1 | 2 - 3 | 350 | 104 | ||
| XM_008765340.1 | XP_008763562.1 | 2 - 3 | 350 | 104 | ||
| GenBank mRNA | AB105879.1 | - | 2 - 3 | 306 | 104 | |
| AF071204.1 | - | 2 - 3 | 141 | 104 | ||
| AF071205.1 | - | 2 - 3 | 142 | 104 | ||
| CB751462.1 | - | 2 - 3 | 306 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kitlg
Genomic Map