Assay Details
| Gene Symbol: Tmem263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241164.2 | XP_006241226.1 | 1 - 2 | 270 | 132 | |
| GenBank mRNA | BC166979.1 | - | 1 - 2 | 140 | 7315264 | 133 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC166979.1,XM_006241164.2 transcripts.
Set Membership: |
|

Tmem263
Genomic Map