Assay Details
| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108083.1 | NP_001101553.1 | 6 - 7 | 2312 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of ubiquitin protein ligase activity
molecular_function
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
biological_process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of ubiquitin protein ligase activity
molecular_function
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P354359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1342 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P354359 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P354359 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P354359 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P354359 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43180 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7816 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10894946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894944 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894945 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894950 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10894942 | - | - |
| Rat Genome 230 2.0 Array | 1382912_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382912_at | - | - |
| Affymetrix HT Rat Focus Array | 1382912_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382912_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700004361.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004363.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004355.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004357.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004356.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004362.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004359.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004360.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004349.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004354.rn.1 | TC0700000520.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004358.rn.1 | TC0700000520.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382912_at | - | - |
| Rat Expression Set 230 | 1382912_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21698574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359606 | - | - |

Usp44
Genomic Map