Assay Details
| Gene Symbol: Glt8d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765296.1 | XP_008763518.1 | 3 - 4 | 775 | 65 | |
| XM_008765297.1 | XP_008763519.1 | 3 - 4 | 774 | 65 | ||
| XM_008765298.1 | XP_008763520.1 | 3 - 4 | 774 | 65 | ||
| XM_008765299.1 | XP_008763521.1 | 3 - 4 | 774 | 65 | ||
More Information
Set Membership: |
|

Glt8d2
Genomic Map