Assay Details
| Gene Symbol: Erich5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241546.2 | XP_006241608.1 | 2 - 3 | 1357 | 147 | |
| XM_006241547.2 | XP_006241609.1 | 2 - 3 | 1348 | 147 | ||
More Information
Set Membership: |
|

Erich5
Genomic Map