Assay Details
| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030841.3 | NP_110468.2 | 4 - 5 | 1377 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P493980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P050030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90015 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P050030 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P493980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P493980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P050030 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P050030 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P493980 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P493980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75640 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10905496 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905494 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905499 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905498 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905493 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905497 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905495 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10905492 | - | - |
| Rat Genome 230 2.0 Array | 1376362_at | - | - |
| Rat Genome 230 2.0 Array | 1387481_at | - | - |
| Rat Genome 230 2.0 Array | 1376362_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387481_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376362_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376362_at | - | - |
| Affymetrix HT Rat Focus Array | 1387481_at | - | - |
| Affymetrix HT Rat Focus Array | 1387481_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700027695.rn.1 | TC0700003512.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376362_PM_at | - | - |
| Rat Expression Set 230 | 1387481_PM_at | - | - |
| Rat Expression Set 230 | 1387481_at | - | - |
| Rat Expression Set 230 | 1376362_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905492 | - | - |
| Rat Genome U34 Set | AF005099_at | - | - |
| Rat Genome U34 Set | rc_AI045501_s_at | - | - |
| Rat Genome U34 Set | rc_AI228113_s_at | - | - |
| Rat Genome U34 Set | rc_AI045501_s_at | - | - |
| Rat Genome U34 Set | rc_AI228113_s_at | - | - |
| Rat Genome U34 Set | AF005099_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21948201 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350450 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52652 | - | - |

Nptxr
Genomic Map