Assay Details
| Gene Symbol: Desi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025703.1 | NP_001020874.1 | 4 - 5 | 469 | 148 | |
| GenBank mRNA | BC098857.1 | - | 4 - 5 | 469 | 7380029 | 148 |
More Information
Updated Mapping Information
Set Membership: |
|

Desi1
Genomic Map