Assay Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134461.1 | NP_604456.1 | 9 - 10 | 1398 | 79 | |
| XM_006229729.2 | XP_006229791.2 | 9 - 10 | 1635 | 79 | ||
| XM_006229730.2 | XP_006229792.1 | 9 - 10 | 1527 | 79 | ||
| XM_006229731.2 | XP_006229793.1 | 9 - 10 | 1771 | 79 | ||
| XM_008759673.1 | XP_008757895.1 | 8 - 9 | 1562 | 79 | ||
| GenBank mRNA | AF484958.1 | - | 9 - 10 | 1398 | 79 | |
| FQ223532.1 | - | 9 - 10 | 1492 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn5
Genomic Map