Assay Details
| Gene Symbol: Nlrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106291.1 | NP_001099761.1 | 1 - 2 | 58 | 138 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106291.1 transcripts.
Set Membership: |
|

Nlrp10
Genomic Map