Assay Details
| Gene Symbol: Cox6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037674.1 | 1 - 2 | 138 | 64 | ||
| GenBank mRNA | BQ782641.1 | - | 2 - 3 | 64 | ||
| FM121690.1 | - | 1 - 2 | 87 | 64 | ||
| FQ217987.1 | - | 1 - 2 | 88 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox6a2
Genomic Map