Assay Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073440.1 | 1 - 2 | 1575 | 92 | ||
| NR_073441.1 | 1 - 2 | 1575 | 92 | |||
| XM_006229688.2 | XP_006229750.1 | 1 - 2 | 1940 | 92 | ||
| XM_008759646.1 | XP_008757868.1 | 1 - 2 | 1940 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crebzf
Genomic Map