Assay Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034911.1 | NP_001030083.1 | 3 - 4 | 577 | 88 | |
| XM_006230189.2 | XP_006230251.1 | 3 - 4 | 850 | 88 | ||
| GenBank mRNA | BC101866.1 | - | 3 - 4 | 577 | 7938969 | 88 |
More Information
Updated Mapping Information
Set Membership: |
|

Ubfd1
Genomic Map