Assay Details
| Gene Symbol: Klhl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229818.1 | XP_006229880.1 | 1 - 2 | 4104 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229818.1 transcripts.
Set Membership: |
|

Klhl35
Genomic Map