Assay Details
| Gene Symbol: Me3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108491.2 | NP_001101961.2 | 6 - 7 | 782 | 76 | |
| XM_006229666.1 | XP_006229728.1 | 6 - 7 | 782 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Me3
Genomic Map